Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGGCTCTTTTCTGCTTTTTAA[C/T]AGGTAAGTTTTTTCTTAAAATTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615868 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPINK6 - serine peptidase inhibitor, Kazal type 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195290.1 | 359 | Missense Mutation | ACA,ATA | T,I 19 | NP_001182219.1 | |
NM_205841.3 | 359 | Missense Mutation | ACA,ATA | T,I 19 | NP_995313.2 |