Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTGTCACTGATAAATGCAATA[G/T]ATACAGGAAGATTTCCACGGTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616704 | ||||||||||||||||||||
Literature Links: |
COMMD10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COMMD10 - COMM domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308080.1 | 537 | Missense Mutation | GAT,TAT | D,Y 12 | NP_001295009.1 | |
NM_016144.3 | 537 | Missense Mutation | GAT,TAT | D,Y 26 | NP_057228.1 | |
XM_011543458.2 | 537 | Missense Mutation | GAT,TAT | D,Y 26 | XP_011541760.1 | |
XM_011543460.2 | 537 | Missense Mutation | GAT,TAT | D,Y 26 | XP_011541762.1 | |
XM_017009553.1 | 537 | Missense Mutation | GAT,TAT | D,Y 12 | XP_016865042.1 | |
XM_017009555.1 | 537 | Missense Mutation | GAT,TAT | D,Y 26 | XP_016865044.1 | |
XM_017009556.1 | 537 | Missense Mutation | GAT,TAT | D,Y 26 | XP_016865045.1 | |
XM_017009557.1 | 537 | Intron | XP_016865046.1 |