Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACTATAATCTGGAGGTTTTAGG[A/T]GTGTGTGGGGTCCTTCCTTTAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610499 | ||||||||||||||||||||
Literature Links: |
RAPGEF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAPGEF6 - Rap guanine nucleotide exchange factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164386.1 | 4783 | Missense Mutation | CAC,CTC | H,L 1534 | NP_001157858.1 | |
NM_001164387.1 | 4783 | Intron | NP_001157859.1 | |||
NM_001164388.1 | 4783 | Intron | NP_001157860.1 | |||
NM_001164389.1 | 4783 | Intron | NP_001157861.1 | |||
NM_001164390.1 | 4783 | Intron | NP_001157862.1 | |||
NM_016340.5 | 4783 | Missense Mutation | CAC,CTC | H,L 1526 | NP_057424.3 |