Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAACGGACAAGAGGCCTGGGTG[C/T]AACGACCAAAATTACCACATCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIFC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIFC1 - kinesin family member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002263.3 | 329 | Missense Mutation | GCA,GTA | A,V 58 | NP_002254.2 | |
XM_011514585.1 | 329 | Missense Mutation | GCA,GTA | A,V 58 | XP_011512887.1 | |
XM_011514587.2 | 329 | Missense Mutation | GCA,GTA | A,V 58 | XP_011512889.1 | |
XM_017010836.1 | 329 | Missense Mutation | GCA,GTA | A,V 58 | XP_016866325.1 | |
XM_017010837.1 | 329 | Missense Mutation | GCA,GTA | A,V 17 | XP_016866326.1 |