Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTATTTCTGAACGGACCTGGGCT[T/C]GTCGGACCAGTGAGCGGCGGCGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615388 MIM: 603164 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAT2 - adenosine deaminase, tRNA specific 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286259.1 | 16 | Intron | NP_001273188.1 | |||
NM_182503.2 | 16 | Intron | NP_872309.2 | |||
XM_011535439.2 | 16 | Intron | XP_011533741.1 | |||
XM_011535440.1 | 16 | Intron | XP_011533742.1 | |||
XM_011535443.1 | 16 | Intron | XP_011533745.1 | |||
XM_011535444.2 | 16 | Intron | XP_011533746.1 | |||
XM_017010260.1 | 16 | Intron | XP_016865749.1 | |||
XM_017010261.1 | 16 | Intron | XP_016865750.1 | |||
XM_017010262.1 | 16 | Intron | XP_016865751.1 | |||
XM_017010263.1 | 16 | Intron | XP_016865752.1 |
PEX3 - peroxisomal biogenesis factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003630.2 | 16 | UTR 5 | NP_003621.1 |