Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGGTGGGAAGAGATTACTGACA[A/G]AACAGACCAAAGTGTTGGGCTTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142855 MIM: 142856 | ||||||||||||||||||||
Literature Links: |
HLA-DMA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HLA-DMA - major histocompatibility complex, class II, DM alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006120.3 | 529 | Missense Mutation | NP_006111.2 |
HLA-DMB - major histocompatibility complex, class II, DM beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |