Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGAGGGCGAGCCTCCACCTGTG[C/G]AGGAGCGGCGGCGGCTGCAGGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 125647 | ||||||||||||||||||||
Literature Links: |
DSP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DSP - desmoplakin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRNP48 - small nuclear ribonucleoprotein U11/U12 subunit 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152551.3 | 84 | Missense Mutation | CAG,GAG | Q,E 9 | NP_689764.3 | |
XM_011514312.2 | 84 | Missense Mutation | CAG,GAG | Q,E 9 | XP_011512614.1 |