Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAATCCAGTCGACGTTCAGGCAC[C/T]GCGCGATGAACGCAAACATGTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600400 | ||||||||||||||||||||
Literature Links: |
PREP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PREP - prolyl endopeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002726.4 | 2338 | Missense Mutation | CAG,CGG | Q,R 702 | NP_002717.3 | |
XM_005267044.2 | 2338 | Intron | XP_005267101.1 | |||
XM_011535925.2 | 2338 | Intron | XP_011534227.1 |