Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGTACGCGGCCCACAGGGAACT[A/G]GAGTCCGGCCCGCGAAGAGCGGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602792 MIM: 602804 MIM: 602811 MIM: 602830 | ||||||||||||||||||||
Literature Links: |
HIST1H2AD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST1H2AD - histone cluster 1, H2ad | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021065.3 | 123 | Nonsense Mutation | CAG,TAG | Q,* 25 | NP_066409.1 |
HIST1H2BF - histone cluster 1, H2bf | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003522.3 | 123 | Intron | NP_003513.1 |
HIST1H3D - histone cluster 1, H3d | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003530.4 | 123 | UTR 5 | NP_003521.2 |
HIST1H4E - histone cluster 1, H4e | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |