Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTATTGGGGGAAAAAAACCACT[A/T]TGAGCACCAAAGCAAGAAAAGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130357 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100130357 - uncharacterized LOC100130357 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D7 - TBC1 domain family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143964.3 | 1130 | UTR 3 | NP_001137436.1 | |||
NM_001143965.3 | 1130 | UTR 3 | NP_001137437.1 | |||
NM_001143966.3 | 1130 | UTR 3 | NP_001137438.1 | |||
NM_001258457.2 | 1130 | UTR 3 | NP_001245386.1 | |||
NM_001318805.1 | 1130 | UTR 3 | NP_001305734.1 | |||
NM_001318806.1 | 1130 | Intron | NP_001305735.1 | |||
NM_016495.5 | 1130 | UTR 3 | NP_057579.1 |
TBC1D7-LOC100130357 - TBC1D7-LOC100130357 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318809.1 | 1130 | Intron | NP_001305738.1 |