Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACGGTCTTTTTGAGGTCAATCA[C/T]GGCGTAGGAGTCTGAGCTTCGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607744 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRS3 - fibroblast growth factor receptor substrate 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006653.4 | 2479 | Missense Mutation | ATG,GTG | M,V 457 | NP_006644.1 | |
XM_011514254.2 | 2479 | Missense Mutation | ATG,GTG | M,V 457 | XP_011512556.1 | |
XM_011514255.2 | 2479 | Missense Mutation | ATG,GTG | M,V 457 | XP_011512557.1 | |
XM_017010193.1 | 2479 | Missense Mutation | ATG,GTG | M,V 420 | XP_016865682.1 |