Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACAGGTTTTCACCAGGTTGGTA[C/G]TGGGGCTCTGTCGCGGTAAAGTAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137025 | ||||||||||||||||||||
Literature Links: |
FYN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FYN - FYN proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002037.5 | 1693 | Missense Mutation | CAC,CAG | H,Q 530 | NP_002028.1 | |
NM_153047.3 | 1693 | Missense Mutation | CAC,CAG | H,Q 527 | NP_694592.1 | |
NM_153048.3 | 1693 | Missense Mutation | CAC,CAG | H,Q 475 | NP_694593.1 | |
XM_005266890.3 | 1693 | Missense Mutation | CAC,CAG | H,Q 527 | XP_005266947.1 | |
XM_005266892.3 | 1693 | Missense Mutation | CAC,CAG | H,Q 475 | XP_005266949.1 | |
XM_017010650.1 | 1693 | Missense Mutation | CAC,CAG | H,Q 530 | XP_016866139.1 | |
XM_017010651.1 | 1693 | Missense Mutation | CAC,CAG | H,Q 530 | XP_016866140.1 | |
XM_017010652.1 | 1693 | Missense Mutation | CAC,CAG | H,Q 530 | XP_016866141.1 | |
XM_017010653.1 | 1693 | Missense Mutation | CAC,CAG | H,Q 530 | XP_016866142.1 | |
XM_017010654.1 | 1693 | Missense Mutation | CAC,CAG | H,Q 530 | XP_016866143.1 | |
XM_017010655.1 | 1693 | Intron | XP_016866144.1 |