Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTGGGCTCTGCGCCCTCATCCTG[C/T]AAGCTCTCCTGGACATGGTAGCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123900 | ||||||||||||||||||||
Literature Links: |
EZR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EZR - ezrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111077.1 | 1648 | Silent Mutation | TTA,TTG | L,L 489 | NP_001104547.1 | |
NM_003379.4 | 1648 | Silent Mutation | TTA,TTG | L,L 489 | NP_003370.2 | |
XM_011536110.1 | 1648 | Silent Mutation | TTA,TTG | L,L 353 | XP_011534412.1 |
MIR3918 - microRNA 3918 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYTL3 - synaptotagmin like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |