Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGGGTGGGGGGTTGGGTTATGA[A/T]GAACTGTGTGAGGAAGGAGAGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601685 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS6KA2 - ribosomal protein S6 kinase A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006932.2 | 4760 | UTR 3 | NP_001006933.2 | |||
NM_001318936.1 | 4760 | UTR 3 | NP_001305865.1 | |||
NM_001318937.1 | 4760 | UTR 3 | NP_001305866.1 | |||
NM_001318938.1 | 4760 | UTR 3 | NP_001305867.1 | |||
NM_021135.5 | 4760 | UTR 3 | NP_066958.2 | |||
XM_006715549.3 | 4760 | UTR 3 | XP_006715612.1 |