Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATTTTATTTCATCCTCTTACCT[C/G]AGCAATCTTGGTTTTCTTTTGTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC101929057 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929057 - uncharacterized LOC101929057 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYRM2 - LYR motif containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDN1 - midasin AAA ATPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014611.2 | 16385 | Missense Mutation | CAG,GAG | Q,E 5465 | NP_055426.1 | |
XM_005248700.3 | 16385 | Missense Mutation | CAG,GAG | Q,E 3460 | XP_005248757.1 | |
XM_006715405.3 | 16385 | Missense Mutation | CAG,GAG | Q,E 3613 | XP_006715468.1 | |
XM_011535635.2 | 16385 | Missense Mutation | CAG,GAG | Q,E 5368 | XP_011533937.1 | |
XM_011535636.2 | 16385 | Intron | XP_011533938.1 |