Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCAGGCTTGGACCTCTTTCAAG[C/G]GGACATCCTCTTGCAGGTGAGTACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600388 | ||||||||||||||||||||
Literature Links: |
MEP1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEP1A - meprin A subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005588.2 | 212 | Missense Mutation | GCG,GGG | A,G 57 | NP_005579.2 | |
XM_011514628.1 | 212 | Missense Mutation | GCG,GGG | A,G 85 | XP_011512930.1 | |
XM_011514629.1 | 212 | Missense Mutation | GCG,GGG | A,G 57 | XP_011512931.1 |