Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAAGCCCAGTGCAATGAAACACA[C/T]GCTCCCAGTCAAACTTCTTGGTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600018 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPCEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPCEF1 - interaction protein for cytohesin exchange factors 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130699.1 | 5211 | UTR 3 | NP_001124171.1 | |||
NM_001130700.1 | 5211 | UTR 3 | NP_001124172.1 | |||
NM_015553.2 | 5211 | UTR 3 | NP_056368.1 | |||
XM_005266919.4 | 5211 | UTR 3 | XP_005266976.1 | |||
XM_005266921.4 | 5211 | UTR 3 | XP_005266978.1 | |||
XM_005266923.3 | 5211 | UTR 3 | XP_005266980.1 | |||
XM_011535742.2 | 5211 | UTR 3 | XP_011534044.1 | |||
XM_011535744.2 | 5211 | UTR 3 | XP_011534046.1 | |||
XM_017010716.1 | 5211 | UTR 3 | XP_016866205.1 | |||
XM_017010717.1 | 5211 | UTR 3 | XP_016866206.1 | |||
XM_017010718.1 | 5211 | UTR 3 | XP_016866207.1 |