Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGAAAGGCCATACTCACACTTT[A/T]ATAGAGATACCAAGTTCTTTAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS10 - mitochondrial ribosomal protein S10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018141.3 | 365 | Silent Mutation | ATA,ATT | I,I 106 | NP_060611.2 | |
XM_005249206.2 | 365 | Silent Mutation | ATA,ATT | I,I 105 | XP_005249263.1 | |
XM_011514724.2 | 365 | Silent Mutation | ATA,ATT | I,I 116 | XP_011513026.1 | |
XM_011514725.2 | 365 | Silent Mutation | ATA,ATT | I,I 115 | XP_011513027.1 | |
XM_017010996.1 | 365 | Silent Mutation | ATA,ATT | I,I 65 | XP_016866485.1 |