Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTGACAGTTTCATGAATAAAGG[C/T]AGTGACAATGAACAAATTCATAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603473 MIM: 608141 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LATS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LATS1 - large tumor suppressor kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC645967 - uncharacterized LOC645967 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP43 - nucleoporin 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198887.2 | 3142 | UTR 3 | NP_942590.1 | |||
XM_005266960.4 | 3142 | UTR 3 | XP_005267017.1 | |||
XM_005266961.3 | 3142 | UTR 3 | XP_005267018.1 | |||
XM_005266962.3 | 3142 | UTR 3 | XP_005267019.1 | |||
XM_011535798.2 | 3142 | Intron | XP_011534100.1 | |||
XM_011535799.2 | 3142 | UTR 3 | XP_011534101.1 |