Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGTGGCTGAGGAGTGGGCCCAG[C/G]GCACCTTCAAACTGAACTCCAATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608310 | ||||||||||||||||||||
Literature Links: |
ASL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASL - argininosuccinate lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000048.3 | 464 | Missense Mutation | CGC,GGC | R,G 77 | NP_000039.2 | |
NM_001024943.1 | 464 | Missense Mutation | CGC,GGC | R,G 77 | NP_001020114.1 | |
NM_001024944.1 | 464 | Missense Mutation | CGC,GGC | R,G 77 | NP_001020115.1 | |
NM_001024946.1 | 464 | Missense Mutation | CGC,GGC | R,G 77 | NP_001020117.1 |