Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACTCGCTGATGCGCTGGGACGC[A/G]TTGAACAAGGTTTCCTCAGAAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 238331 MIM: 150240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLD - dihydrolipoamide dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAMB1 - laminin subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002291.2 | 5264 | Silent Mutation | AAC,AAT | N,N 1643 | NP_002282.2 | |
XM_017012201.1 | 5264 | Silent Mutation | AAC,AAT | N,N 1667 | XP_016867690.1 | |
XM_017012202.1 | 5264 | Intron | XP_016867691.1 |