Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACGTACTTGCCATTGAACACAGA[A/G]TTGTTGATCCAACGCCACCTTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604987 | ||||||||||||||||||||
Literature Links: |
CLEC5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC5A - C-type lectin domain family 5 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301167.1 | 519 | Silent Mutation | AAC,AAT | N,N 122 | NP_001288096.1 | |
NM_013252.2 | 519 | Silent Mutation | AAC,AAT | N,N 145 | NP_037384.1 | |
XM_011515995.2 | 519 | Silent Mutation | AAC,AAT | N,N 102 | XP_011514297.1 | |
XM_017011915.1 | 519 | Silent Mutation | AAC,AAT | N,N 125 | XP_016867404.1 | |
XM_017011916.1 | 519 | Silent Mutation | AAC,AAT | N,N 145 | XP_016867405.1 | |
XM_017011917.1 | 519 | Silent Mutation | AAC,AAT | N,N 122 | XP_016867406.1 |