Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATACTGCCTTCAGAAAAGCAATC[C/T]CTGGTCCACAAATTATTCATGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614965 MIM: 612432 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCRN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCRN1 - secernin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145513.1 | 4065 | UTR 3 | NP_001138985.1 | |||
NM_001145514.1 | 4065 | UTR 3 | NP_001138986.1 | |||
NM_001145515.1 | 4065 | UTR 3 | NP_001138987.1 | |||
NM_014766.4 | 4065 | UTR 3 | NP_055581.3 |
WIPF3 - WAS/WASL interacting protein family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |