Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGTGAGACAAAGTCTTTACTTC[C/T]AGGGTTTGCCTTGCAGGTGACCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610523 | ||||||||||||||||||||
Literature Links: |
CEP41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP41 - centrosomal protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257158.1 | 1320 | Nonsense Mutation | TAG,TGG | *,W 300 | NP_001244087.1 | |
NM_001257159.1 | 1320 | Nonsense Mutation | TAG,TGG | *,W 284 | NP_001244088.1 | |
NM_001257160.1 | 1320 | Intron | NP_001244089.1 | |||
NM_018718.2 | 1320 | Nonsense Mutation | TAG,TGG | *,W 372 | NP_061188.1 | |
XM_011516709.2 | 1320 | Nonsense Mutation | TAG,TGG | *,W 337 | XP_011515011.1 | |
XM_011516710.2 | 1320 | Nonsense Mutation | TAG,TGG | *,W 337 | XP_011515012.1 |