Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAACGTGACGGCGACCGAGGGGC[A/G]GAATGTGGAGATGTCCTGCGCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
VSTM2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VSTM2A - V-set and transmembrane domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301009.1 | 531 | Missense Mutation | CAG,CGG | Q,R 42 | NP_001287938.1 | |
NM_001317843.1 | 531 | Missense Mutation | CAG,CGG | Q,R 42 | NP_001304772.1 | |
NM_182546.3 | 531 | Missense Mutation | CAG,CGG | Q,R 42 | NP_872352.2 | |
XM_006715665.3 | 531 | Missense Mutation | CAG,CGG | Q,R 42 | XP_006715728.1 | |
XM_006715666.3 | 531 | Missense Mutation | CAG,CGG | Q,R 42 | XP_006715729.1 | |
XM_017011841.1 | 531 | Missense Mutation | CAG,CGG | Q,R 42 | XP_016867330.1 |