Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGGCGCTGGTCGGCAGTCCGAA[A/C]GGAGGGAGCGAGACCCCCAAGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142996 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EVX1 - even-skipped homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304519.1 | 948 | UTR 5 | NP_001291448.1 | |||
NM_001304520.1 | 948 | UTR 5 | NP_001291449.1 | |||
NM_001989.4 | 948 | Missense Mutation | AAA,AAC | K,N 154 | NP_001980.1 |
EVX1-AS - EVX1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |