Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCGGAGGTAGCCAGCTCCCAGC[A/G]GTCCACAGGCGGTCCACAGGCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602909 MIM: 612546 | ||||||||||||||||||||
Literature Links: |
CLDN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLDN4 - claudin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBSCR27 - Williams Beuren syndrome chromosome region 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152559.2 | 738 | Silent Mutation | ACC,ACT | T,T 206 | NP_689772.2 | |
XM_017011777.1 | 738 | Silent Mutation | ACC,ACT | T,T 232 | XP_016867266.1 | |
XM_017011778.1 | 738 | Silent Mutation | ACC,ACT | T,T 232 | XP_016867267.1 |