Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTGAGGACAGAGATCCTTGGGTG[G/T]CGGGAGGCGTGGCTGAGCACTGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610486 MIM: 602181 | ||||||||||||||||||||
Literature Links: |
LRRC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC4 - leucine rich repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022143.4 | 1634 | Silent Mutation | CGA,CGC | R,R 397 | NP_071426.1 | |
XM_011516461.2 | 1634 | Silent Mutation | CGA,CGC | R,R 397 | XP_011514763.1 | |
XM_017012505.1 | 1634 | Silent Mutation | CGA,CGC | R,R 397 | XP_016867994.1 |
SND1 - staphylococcal nuclease and tudor domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017011987.1 | 1634 | Intron | XP_016867476.1 |