Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 | ||||||||||||||||||||
Literature Links: |
AP4M1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP4M1 - adaptor related protein complex 4 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COPS6 - COP9 signalosome subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006833.4 | 2693 | Intron | NP_006824.2 |
MCM7 - minichromosome maintenance complex component 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278595.1 | 2693 | Missense Mutation | CGC,TGC | R,C 507 | NP_001265524.1 | |
NM_005916.4 | 2693 | Missense Mutation | CGC,TGC | R,C 683 | NP_005907.3 | |
NM_182776.2 | 2693 | Missense Mutation | CGC,TGC | R,C 507 | NP_877577.1 | |
XM_005250348.3 | 2693 | Missense Mutation | CGC,TGC | R,C 576 | XP_005250405.1 | |
XM_017012217.1 | 2693 | Missense Mutation | CGC,TGC | R,C 576 | XP_016867706.1 |
MIR106B - microRNA 106b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR25 - microRNA 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR93 - microRNA 93 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |