Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGCGGAGCTGCAGAGATGCAA[A/G]GCCAGGCTACACGCCATGGAGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612545 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD11 - abhydrolase domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145364.2 | Intron | NP_001138836.1 | ||||
NM_001301058.1 | Intron | NP_001287987.1 | ||||
NM_001321382.1 | Intron | NP_001308311.1 | ||||
NM_001321383.1 | Intron | NP_001308312.1 | ||||
NM_148912.3 | Intron | NP_683710.1 | ||||
NM_148913.3 | Intron | NP_683711.1 | ||||
XM_006716144.2 | Intron | XP_006716207.1 | ||||
XM_011516590.2 | Intron | XP_011514892.1 | ||||
XM_011516591.1 | Intron | XP_011514893.1 |
ABHD11-AS1 - ABHD11 antisense RNA 1 (tail to tail) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |