Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACCAGCCCAGGAAGCAGCAGGG[A/T]CACCAAGGGTCCAGTTTCAGTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM200A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM200A - family with sequence similarity 200 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF655 - zinc finger protein 655 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009958.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_001009958.1 | |
NM_001009960.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_001009960.1 | |
NM_001083956.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_001077425.1 | |
NM_001085366.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_001078835.1 | |
NM_001085367.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_001078836.1 | |
NM_001085368.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_001078837.1 | |
NM_024061.3 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_076966.1 | |
NM_138494.2 | 271 | Missense Mutation | ACA,TCA | T,S 12 | NP_612503.1 | |
XM_017012598.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868087.1 | |
XM_017012599.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868088.1 | |
XM_017012600.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868089.1 | |
XM_017012601.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868090.1 | |
XM_017012602.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868091.1 | |
XM_017012603.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868092.1 | |
XM_017012604.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868093.1 | |
XM_017012605.1 | 271 | UTR 5 | XP_016868094.1 | |||
XM_017012606.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868095.1 | |
XM_017012607.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868096.1 | |
XM_017012608.1 | 271 | UTR 5 | XP_016868097.1 | |||
XM_017012609.1 | 271 | UTR 5 | XP_016868098.1 | |||
XM_017012610.1 | 271 | UTR 5 | XP_016868099.1 | |||
XM_017012611.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868100.1 | |
XM_017012612.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868101.1 | |
XM_017012613.1 | 271 | Missense Mutation | ACA,TCA | T,S 12 | XP_016868102.1 |