Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCTCCCAGGAGCACACCCCGCA[C/T]CCCTGCCGCAGCGCCATGGCCCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615477 MIM: 611914 | ||||||||||||||||||||
Literature Links: |
NYAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NYAP1 - neuronal tyrosine phosphorylated phosphoinositide-3-kinase adaptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173564.3 | 430 | Silent Mutation | CAC,CAT | H,H 72 | NP_775835.2 | |
XM_006715907.3 | 430 | Silent Mutation | CAC,CAT | H,H 72 | XP_006715970.1 | |
XM_017011868.1 | 430 | Silent Mutation | CAC,CAT | H,H 72 | XP_016867357.1 |
TSC22D4 - TSC22 domain family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |