Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGCGGACTTCGACACCTACGAC[A/G]ATCGGGCCTACAGCAGCTTCGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603431 | ||||||||||||||||||||
Literature Links: |
EIF4H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF4H - eukaryotic translation initiation factor 4H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022170.1 | Intron | NP_071496.1 | ||||
NM_031992.1 | Intron | NP_114381.1 | ||||
XM_011516556.1 | Intron | XP_011514858.1 | ||||
XM_011516557.1 | Intron | XP_011514859.1 |