Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATTGTTAAGAAAAATATTGAGC[A/T]TTTCACTTCACCATCAAAAAATTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606872 MIM: 126650 | ||||||||||||||||||||
Literature Links: |
CBLL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBLL1 - Cbl proto-oncogene like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC26A3 - solute carrier family 26 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000111.2 | 2327 | Missense Mutation | AGC,TGC | S,C 706 | NP_000102.1 | |
XM_011515867.2 | 2327 | Missense Mutation | AGC,TGC | S,C 706 | XP_011514169.1 |