Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCACACACCAGCAGAAGCCCCG[C/T]TTCCTGCCTTTGGAAGGGCGACACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146732 | ||||||||||||||||||||
Literature Links: |
IGFBP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGFBP3 - insulin like growth factor binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000598.4 | 924 | Silent Mutation | AAA,AAG | K,K 258 | NP_000589.2 | |
NM_001013398.1 | 924 | Silent Mutation | AAA,AAG | K,K 264 | NP_001013416.1 | |
XM_017012152.1 | 924 | Silent Mutation | AAA,AAG | K,K 258 | XP_016867641.1 |