Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAATCCTCGTGGCCTTGGCAGT[A/C]TTTTTTCTTGTCTCCACTCAGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 162320 | ||||||||||||||||||||
Literature Links: |
TAC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAC1 - tachykinin precursor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003182.2 | 273 | Silent Mutation | GTA,GTC | V,V 9 | NP_003173.1 | |
NM_013996.2 | 273 | Silent Mutation | GTA,GTC | V,V 9 | NP_054702.1 | |
NM_013997.2 | 273 | Silent Mutation | GTA,GTC | V,V 9 | NP_054703.1 | |
NM_013998.2 | 273 | Silent Mutation | GTA,GTC | V,V 9 | NP_054704.1 |