Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCAAGTTAGTTAAGAGAAATTT[C/T]CTTGAGGCGCTAAAGTCCAATGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605761 | ||||||||||||||||||||
Literature Links: |
ASB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASB4 - ankyrin repeat and SOCS box containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016116.2 | 134 | Silent Mutation | TTC,TTT | F,F 21 | NP_057200.1 | |
NM_145872.2 | 134 | Silent Mutation | TTC,TTT | F,F 21 | NP_665879.1 | |
XM_017012303.1 | 134 | Silent Mutation | TTC,TTT | F,F 21 | XP_016867792.1 |