Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTGGGAAGGTAGTTTATCCAAC[A/G]TAAGGATCCCATCCTTTTCCATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPVL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPVL - carboxypeptidase, vitellogenic like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019029.2 | 1268 | Silent Mutation | TAC,TAT | Y,Y 474 | NP_061902.2 | |
NM_031311.3 | 1268 | Silent Mutation | TAC,TAT | Y,Y 474 | NP_112601.3 | |
XM_011515437.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 474 | XP_011513739.1 | |
XM_017012366.1 | 1268 | Silent Mutation | TAC,TAT | Y,Y 383 | XP_016867855.1 |
LOC100506497 - uncharacterized LOC100506497 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |