Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTGTTCACCTTCTGTTTCTGAC[A/G]CATCCAGTGATTCTTCAGGTTCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614903 | ||||||||||||||||||||
Literature Links: |
SNX16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX16 - sorting nexin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022133.3 | 1115 | Missense Mutation | GCG,GTG | A,V 285 | NP_071416.2 | |
NM_152836.2 | 1115 | Missense Mutation | GCG,GTG | A,V 285 | NP_690049.1 | |
NM_152837.2 | 1115 | Missense Mutation | GCG,GTG | A,V 256 | NP_690050.1 | |
XM_005251282.4 | 1115 | Missense Mutation | GCG,GTG | A,V 256 | XP_005251339.1 | |
XM_005251283.2 | 1115 | Missense Mutation | GCG,GTG | A,V 256 | XP_005251340.1 | |
XM_011517574.2 | 1115 | Missense Mutation | GCG,GTG | A,V 128 | XP_011515876.1 |