Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATACTGGCTGCTGTCCGAGCAGC[C/T]CTAGTGACTGTCTTGGGGGCAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYN - LYN proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111097.2 | Intron | NP_001104567.1 | ||||
NM_002350.3 | Intron | NP_002341.1 | ||||
XM_011517529.2 | Intron | XP_011515831.1 | ||||
XM_017013415.1 | Intron | XP_016868904.1 | ||||
XM_017013416.1 | Intron | XP_016868905.1 |