Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCACTTTATCCATACAATACTT[C/T]AGTCTTTTAAGAGAATACTGAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF250 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF250 - zinc finger protein 250 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109689.3 | 2261 | UTR 3 | NP_001103159.1 | |||
NM_021061.4 | 2261 | UTR 3 | NP_066405.1 | |||
XM_005272327.1 | 2261 | UTR 3 | XP_005272384.1 | |||
XM_005272328.3 | 2261 | UTR 3 | XP_005272385.1 | |||
XM_006716612.3 | 2261 | UTR 3 | XP_006716675.1 | |||
XM_006716613.3 | 2261 | UTR 3 | XP_006716676.1 | |||
XM_011517209.1 | 2261 | UTR 3 | XP_011515511.1 |