Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATCTTCAGAATGAGTCAGCTCC[A/G]CTTGTCCCAGGCAGGAGACTCACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607837 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLN8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLN8 - ceroid-lipofuscinosis, neuronal 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018941.3 | 47 | Intron | NP_061764.2 | |||
XM_005266021.3 | 47 | Intron | XP_005266078.1 | |||
XM_005266022.1 | 47 | UTR 5 | XP_005266079.1 | |||
XM_005266023.1 | 47 | Intron | XP_005266080.1 | |||
XM_011534745.1 | 47 | Intron | XP_011533047.1 | |||
XM_011534746.2 | 47 | Intron | XP_011533048.1 | |||
XM_011534747.1 | 47 | Intron | XP_011533049.1 |
LOC101927752 - uncharacterized LOC101927752 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |