Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGTACTTCACAAGGATAGAGTG[A/G]AGTTGCCTGGCCCCCGACCAGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF517 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF517 - zinc finger protein 517 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317936.1 | 730 | Nonsense Mutation | TGA,TGG | *,W 28 | NP_001304865.1 | |
NM_213605.2 | 730 | Nonsense Mutation | TGA,TGG | *,W 28 | NP_998770.2 | |
XM_011517014.2 | 730 | Nonsense Mutation | TGA,TGG | *,W 28 | XP_011515316.1 | |
XM_011517015.2 | 730 | Nonsense Mutation | TGA,TGG | *,W 28 | XP_011515317.1 | |
XM_011517017.2 | 730 | Intron | XP_011515319.1 | |||
XM_017013384.1 | 730 | Intron | XP_016868873.1 |