Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACAGTTATTTGGCTCAAATTCC[C/T]GTGATCACATATTCAGAAAACTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611238 MIM: 603026 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHCHD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHCHD7 - coiled-coil-helix-coiled-coil-helix domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011667.2 | Intron | NP_001011667.1 | ||||
NM_001011668.2 | Intron | NP_001011668.1 | ||||
NM_001011669.2 | Intron | NP_001011669.1 | ||||
NM_001011670.2 | Intron | NP_001011670.1 | ||||
NM_001011671.2 | Intron | NP_001011671.1 | ||||
NM_001317858.1 | Intron | NP_001304787.1 | ||||
NM_001317859.1 | Intron | NP_001304788.1 | ||||
NM_024300.4 | Intron | NP_077276.2 |
PLAG1 - PLAG1 zinc finger | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |