Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGGTGTTTGATCAACTTTGAT[C/T]GCTGGCTGAAGGCTTTCCCACAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601262 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF16 - zinc finger protein 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029976.2 | 2192 | Missense Mutation | CAA,CGA | Q,R 669 | NP_001025147.2 | |
NM_006958.2 | 2192 | Missense Mutation | CAA,CGA | Q,R 669 | NP_008889.2 | |
XM_005272341.2 | 2192 | Missense Mutation | CAA,CGA | Q,R 669 | XP_005272398.1 | |
XM_005272342.2 | 2192 | Missense Mutation | CAA,CGA | Q,R 529 | XP_005272399.1 | |
XM_011517298.1 | 2192 | Missense Mutation | CAA,CGA | Q,R 669 | XP_011515600.1 |