Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACTGGACAAGTCCCAGGACAAC[C/T]GCGGCCAGGGAGTACTGCCCAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY8 - adenylate cyclase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115.2 | 3967 | Silent Mutation | GCA,GCG | A,A 1194 | NP_001106.1 | |
XM_005250769.3 | 3967 | Silent Mutation | GCA,GCG | A,A 1164 | XP_005250826.1 | |
XM_006716501.3 | 3967 | Silent Mutation | GCA,GCG | A,A 1128 | XP_006716564.1 | |
XM_017013006.1 | 3967 | Silent Mutation | GCA,GCG | A,A 1098 | XP_016868495.1 | |
XM_017013007.1 | 3967 | Intron | XP_016868496.1 |