Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAGGGCATGGACTCACACGAAC[A/G]ACGCGGATGCTGCTGCGGGCAGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138251 MIM: 609564 MIM: 601282 | ||||||||||||||||||||
Literature Links: |
GRINA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRINA - glutamate ionotropic receptor NMDA type subunit associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARP10 - poly(ADP-ribose) polymerase family member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317895.1 | 2662 | Silent Mutation | GTC,GTT | V,V 862 | NP_001304824.1 | |
NM_032789.4 | 2662 | Silent Mutation | GTC,GTT | V,V 850 | NP_116178.2 | |
XM_011517336.2 | 2662 | Silent Mutation | GTC,GTT | V,V 841 | XP_011515638.1 |
PLEC - plectin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |