Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGCTCCATGAAGGAGACTTC[C/G]GAGTACCTGGCATGGAAGTGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615563 MIM: 604819 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6845 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6845 - microRNA 6845 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRBP2 - nuclear receptor binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178564.3 | 1223 | Silent Mutation | TCC,TCG | S,S 361 | NP_848659.2 | |
XM_017013378.1 | 1223 | Silent Mutation | TCC,TCG | S,S 361 | XP_016868867.1 | |
XM_017013379.1 | 1223 | Silent Mutation | TCC,TCG | S,S 361 | XP_016868868.1 | |
XM_017013380.1 | 1223 | Silent Mutation | TCC,TCG | S,S 361 | XP_016868869.1 | |
XM_017013381.1 | 1223 | Silent Mutation | TCC,TCG | S,S 361 | XP_016868870.1 | |
XM_017013382.1 | 1223 | Silent Mutation | TCC,TCG | S,S 361 | XP_016868871.1 | |
XM_017013383.1 | 1223 | Silent Mutation | TCC,TCG | S,S 361 | XP_016868872.1 |
PUF60 - poly(U) binding splicing factor 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |