Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCCCTCAGGCATCTGGATCCAT[A/G]GGAGCACCCTCCGAAGCCCGTGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606110 | ||||||||||||||||||||
Literature Links: |
LYNX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LYNX1 - Ly6/neurotoxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023946.3 | 128 | Missense Mutation | CAT,TAT | H,Y 70 | NP_076435.1 | |
NM_177457.4 | 128 | Intron | NP_803252.1 | |||
NM_177458.1 | 128 | Missense Mutation | CAT,TAT | H,Y 36 | NP_803253.1 | |
NM_177476.3 | 128 | Intron | NP_803429.1 | |||
NM_177477.3 | 128 | Intron | NP_803430.1 |