Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTGATAAAACATCTGTAGAAAA[G/T]TTAACTGTCTTGCCAAACCCATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB1 - EF-hand calcium binding domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142857.1 | 1120 | Intron | NP_001136329.1 | |||
NM_024593.3 | 1120 | UTR 3 | NP_078869.1 | |||
XM_005251303.1 | 1120 | UTR 3 | XP_005251360.1 | |||
XM_011517589.1 | 1120 | Intron | XP_011515891.1 | |||
XM_011517590.2 | 1120 | UTR 3 | XP_011515892.1 | |||
XM_011517591.2 | 1120 | UTR 3 | XP_011515893.1 | |||
XM_017013826.1 | 1120 | UTR 3 | XP_016869315.1 | |||
XM_017013827.1 | 1120 | UTR 3 | XP_016869316.1 | |||
XM_017013828.1 | 1120 | Intron | XP_016869317.1 |